missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Hydroxychloroquine Sulfate

Catalog No. 5092720001
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5092720001 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5092720001 Supplier MilliporeSigma™ Supplier No. 5.09272.0001

A cell-permeable compound that increases intralysosomal pH and impairs autophagic degradation process. Suppresses proliferation of human dermal fibroblasts (IC50 = 30 μM).

  • Packaged Under Inert Gas
  • Deactivate with 5% Bleach

Specifications

Color Off-white
Description 98% by HPLC
Molecular Formula C18H28ClN3O5S
Solubility H2O
Content And Storage Protect from light
Form Powder
Quantity 25 mg