missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Human IgM, Plasma

Numéro de catalogue. 4017991MG
Modifier l'affichage
Click to view available options
Quantité:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
4017991MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 4017991MG Fournisseur MilliporeSigma™ Code fournisseur 4017991MG

Native IgM from human plasma

Spécifications

Formule In 200mM NaCl, 50mM Tris, pH 8.0
Quantité 1 mg
Spécifications de test Nonreactive for HBsAg and for HCV, HBc, HIV-1, and HIV-2 antibodies
Contenu et stockage -20°C
Forme Liquid

For Research Use Only