missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Human γ-Globulin, Serum

Catalog No. 3458865GM
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3458865GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3458865GM Supplier MilliporeSigma™ Supplier No. 3458865GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Temporarily boosts patient's immunity against disease

Specifications

Quantity 5 g
Content And Storage 2°C to 8°C
Form Lyophilized
Purity or Quality Grade ≥90% by electrophoresis
Specificity Negative for HBsAg and for HIV and HCV antibodies

For Research Use Only