missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ HSP70 Inhibitor, PES-Cl

Catalog No. 5310670001
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5310670001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5310670001 Supplier MilliporeSigma™ Supplier No. 5.31067.0001

Directly binds to HSP70 and disrupts its interaction with client proteins..

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach

Specifications

Color White
Description 98% by HPLC
Target Proteins HSP70
Molecular Formula C8H6ClNO2S
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg