missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Horseradish Peroxidase
Suitable for conjugation to proteins and antibodies and as cytochemical marker in immunohistochemical staining
Specifications
| Content And Storage | -20°C |
|---|---|
| Product Type | Horseradish Peroxidase |
| Form | Lyophilized Solid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
5165315KU
|
MilliporeSigma™
5165315KU |
5 kU |
N/A
|
|
|||||
|
51653125KU
|
MilliporeSigma™
51653125KU |
25 kU |
N/A
|
|
|||||
Description
Note: 1KU = 1000 units.
Specifications
| -20°C | |
| Horseradish Peroxidase | |
| Lyophilized Solid |
For Research Use Only