missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Horseradish Peroxidase

Catalog No. 5165315KU
Change view
Click to view available options
Quantity:
25 kU
5 kU
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5165315KU 5 kU
51653125KU 25 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5165315KU Supplier MilliporeSigma™ Supplier No. 5165315KU

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Suitable for conjugation to proteins and antibodies and as cytochemical marker in immunohistochemical staining

Note: 1KU = 1000 units.

Specifications

Content And Storage -20°C
Quantity 5 kU
Product Type Horseradish Peroxidase
Form Lyophilized Solid

For Research Use Only