missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Highly Purified Protease-Free RNase A, Bovine Pancreas
For selective degradation of RNA
$191.00 - $756.00
Specifications
| Content And Storage | -20°C |
|---|---|
| Source | Bovine pancreas |
| Product Type | Protease-Free RNase A |
| Form | Lyophilized Solid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
55674610KU
|
MilliporeSigma™
55674610KU |
10 kU |
Each for $191.00
|
|
|||||
|
55674650KU
|
MilliporeSigma™
55674650KU |
50 kU |
Each for $756.00
|
|
|||||
Description
- Chromatographically purified
- Pyrimidine-specific endoribonuclease that acts on single-stranded RNA
Specifications
| -20°C | |
| Bovine pancreas | |
| Protease-Free RNase A | |
| Lyophilized Solid |
For Research Use Only