missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ HIF-Hydroxylase Inhibitor, DMOG

Catalog No. 40009150MG
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40009150MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40009150MG Supplier MilliporeSigma™ Supplier No. 40009150MG

The HIF-Hydroxylase Inhibitor, DMOG, also referenced under CAS 89464-63-1, controls the biological activity of HIF-Hydroxylase. This small molecule/inhibitor is primarily used for Cell Structure applications.

A cell-permeable 2-OG (2-oxoglutarate) analog that is expected to act as a competitive inhibitor against all 2-OG-dependent dioxygenases. Effectively upregulates normoxia HIF-α transcription activity (effective conc. = 1mm in cultures) by inhibiting HIF-PH- (HIF-α-prolyl hydroxylase) catalyzed ODD (oxygen-dependent degradation domain) Prolyl hydroxylation (Pro564 and Pro530 in HIF-1α and HIF-2α, respectively) and aspartyl/asparanginyl β-hydroxylase-catalyzed CAD Asp hydroxylation (Asn803 and Asn851 in HIF-1α and HIF-2α, respectively), resulting in a blockage of pVHL (von Hippel-Lindau) ubiquitin ligase complex-mediated HIF-α degradation and a simultaneous promotion of HIF CAD (COOH-terminal activation domain) p300/CBP association. DMOG is also demonstrated to augment artery ligation-induced up-regulation of HIF-1α and FLK (by 100% and 40%, respectively, in 11 days; 8mg/0.5 ml/animal/i.p.; q.o.d.) in muscle tissues in a murine ischaemic model in vivo .

Specifications

Color White
Description 98% by HPLC
CAS 89464-63-1
Molecular Formula C6H9NO5
Formula Weight 175.1g/mol
Solubility DMSO
Content And Storage Hygroscopic
Form Solid
Quantity 50 mg