missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Granzyme B Inhibitor I

Catalog No. 3680501MG
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3680501MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3680501MG Supplier MilliporeSigma™ Supplier No. 3680501MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Granzyme B Inhibitor I controls the biological activity of Granzyme B. This small molecule/inhibitor is primarily used for Cancer applications.

  • Peptide Sequence: Z-Ala-Ala-Asp-CH2Cl
  • Cell Permeable

Specifications

Color Off-white
Description 95% by HPLC
Product Type Inhibitor
Target Proteins Granzyme B
Molecular Formula C19H24ClN3O7
Formula Weight 441.9g/mol
Solubility DMSO
Content And Storage Protect from moisture
Form Solid
Quantity 1 mg