missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Glyburide

Catalog No. 3563101GM
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3563101GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3563101GM Supplier MilliporeSigma™ Supplier No. 3563101GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A sulfonylurea that selectively blocks ATP-sensitive K+ channels.

A sulfonylurea that selectively blocks ATP-sensitive K+ channels. Reduced K+ conductance causes membrane depolarization and influx of Ca2+ through voltage sensitive Ca2+ channels. Blocks vascular smooth muscle relaxation produced by KATP channel openers. Inhibits the vasorelaxing effects of nitric oxide.

Specifications

Color White
Description 98% by TLC
Product Type Inhibitor
Target Proteins ATP-sensitive K+ channels
CAS 10238-21-8
Molecular Formula C23H28ClN3O5S
Formula Weight 494g/mol
Solubility DMSO or EtOH (slightly)
Form Solid
Quantity 1 g