missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Glucose Transporter Inhibitor IV, WZB117

Catalog No. 40003625MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40003625MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40003625MG Supplier MilliporeSigma™ Supplier No. 40003625MG

The Glucose Transporter Inhibitor IV, WZB117 controls the biological activity of Glucose Transporter.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach

Specifications

Color White
Description 98% by HPLC
Target Proteins Glut1
CAS 1223397-11-2
Formula Weight 368.3g/mol
Solubility EtOH
Content And Storage Protect from Light, Protect from Moisture
Form Powder
Quantity 25 mg