missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Furin Inhibitor I
Description
Peptide Sequence: Decanoyl-Arg-Val-Lys-Arg-CMK
Specifications
Specifications
| Color | Off-white |
| Formulation | Supplied as a trifluoroacetate salt. |
| Description | 90% by HPLC |
| Target Proteins | viral glycoproteins |
| Molecular Formula | C34H66ClN11O5 |
| Formula Weight | 744.4g/mol |
| Solubility | DMSO |
| Content And Storage | Protect from moisture |
| Form | Solid |
| Quantity | 1 mg |