missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Fumitremorgin C, Aspergillus. fumigatus

Catalog No. 344847250UG
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
344847250UG 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 344847250UG Supplier MilliporeSigma™ Supplier No. 344847250UG

A cell-permeable indolyldiketopiperazinyl mycotoxin that inhibits BCRP/ABCG2 (breast cancer resistance protein/ATP-binding cassette G2) multidrug transport activity.

  • Packaged Under Inert Gas
  • Cell Permeable

Specifications

Description 95% by HPLC
Target Proteins BCRP/ABCG2 (breast cancer resistance protein/ATP-binding cassette G2)
CAS 118974-02-0
Molecular Formula C22H25N3O3
Formula Weight 379.5g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Lyophilized
Quantity 250 μg