missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Fucosyltransferase Inhibitor, 2F-Peracetyl-Fucose
2F-Peracetyl-Fucose is a cell-permeable fluorinated fucose derivative that acts as an inhibitor of fucosyltransferases following its uptake and metabolic transformation into a GDP-fucose mimetic.
Supplier: MilliporeSigma™ 34482710MG
Description
- Packaged Under Inert Gas
- Cell Permeable
- Deactivate with 5% Bleach
Specifications
| White | |
| Fucosyltransferase | |
| 292.3g/mol | |
| Protect from light | |
| 10 mg |
| 98% by NMR | |
| C12H17FO7 | |
| DMSO | |
| Solid |