missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Forskolin, Coleus forskohlii in DMSO

Catalog No. 3442825MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3442825MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3442825MG Supplier MilliporeSigma™ Supplier No. 3442825MG

The major cell-permeable diterpene isolated from the Indian plant Coleus forskohlii.

Packaged Under Inert Gas

Specifications

Formulation A 50 mM (5mg/244μl) solution of Forskolin, Coleus forskohlii in DMSO.
Description 97% by HPLC
Molecular Formula C22H34O7
Formula Weight 410.5g/mol
Content And Storage Hygroscopic, Protect from light
Form Liquid
Quantity 5 mg