missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Ezrin Inhibitor, NSC668394

Catalog No. 34121610MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34121610MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34121610MG Supplier MilliporeSigma™ Supplier No. 34121610MG

The Ezrin Inhibitor, NSC668394 controls the biological activity of Ezrin.

Packaged Under Inert Gas

Specifications

Color Reddish Orange
Description 95% by HPLC
Molecular Formula C17H12Br2N2O3
Formula Weight 449.9g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg