missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Erastin

Catalog No. 3296005MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3296005MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3296005MG Supplier MilliporeSigma™ Supplier No. 3296005MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A cell-permeable piperazinyl-quinazolinone compound that exhibits oncogene-selective lethality.

Packaged Under Inert Gas

Specifications

Color White
Description 98% by HPLC
CAS 571203-78-6
Molecular Formula C30H31ClN4O4
Formula Weight 547.1g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg