missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Epoxomicin, Synthetic

Catalog No. 324800100UG
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
324800100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 324800100UG Supplier MilliporeSigma™ Supplier No. 324800100UG

Epoxomicin, Synthetic, CAS 134381-21-8, is a potent, specific, and irreversible inhibitor of chymotrypsin-like, trypsin-like, and peptidyl-glutamyl peptide hydrolyzing activities of the proteasome.

Packaged Under Inert Gas

Specifications

Color White
Description 95% by HPLC
Target Proteins CT-L,T-L,PGPH activity of proteasome
Molecular Formula C28H50N4O7
Formula Weight 554.7g/mol
Solubility DMSO or MeOH
Content And Storage Protect from light
Form Solid
Quantity 100 μg