missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ EPAC Inhibitor, ESI-09

Catalog No. 5005060001
Change view
Click to view available options
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
5005060001 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5005060001 Supplier MilliporeSigma™ Supplier No. 5.00506.0001

A cell-permeable compound that targets cAMP-binding domain of EPAC/cAMP-GEF and inhibits cAMP-dependent EPAC GEF activity (IC50 = 1.4 and 3.2 μM, respectively, against EPAC2 and EPAC1).

Packaged Under Inert Gas

Specifications

Color Yellow
Description 98% by HPLC
Molecular Formula C16H15ClN4O2
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg