missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ eIF2-GTP-tRNAiMet ternary complex Inhibitor Negative Control, NCPdCPU

Catalog No. 32488910MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32488910MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32488910MG Supplier MilliporeSigma™ Supplier No. 32488910MG

The eIF2-GTP-tRNAiMet ternary complex Inhibitor Negative Control, NCPdCPU controls the biological activity of eIF2-GTP-tRNAiMet ternary complex.

Packaged Under Inert Gas

Specifications

Color Yellow
Description 99% by HPLC
Molecular Formula C13H8Cl3N3O3
Formula Weight 360.58g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg