missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Dynamin Inhibitor I, Dynasore

Catalog No. 32441010MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32441010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32441010MG Supplier MilliporeSigma™ Supplier No. 32441010MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Dynamin Inhibitor I, Dynasore, also referenced under CAS 304448-55-3, controls the biological activity of Dynamin. This small molecule/inhibitor is primarily used for Membrane applications.

Packaged Under Inert Gas

Specifications

Color Pale Yellow
Description 98% by HPLC
Product Type Inhibitor
CAS 304448-55-3
Molecular Formula C18H14N2O4
Formula Weight 322.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg