missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Dexamethasone

Catalog No. 265005100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
265005100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 265005100MG Supplier MilliporeSigma™ Supplier No. 265005100MG

Most active and highly stable glucocorticoid.

Most active and highly stable glucocorticoid. Causes a reduction in cyclin A and Cdk2 activity, and inhibition of G1/S transition in osteoblasts. Inhibits the phosphorylation of retinoblastoma (Rb) protein. Inhibits the expression of inducible but not constitutive nitric oxide synthase in vascular endothelial cells (IC50 = 5 nM). Enhances active cation transport in aortic smooth muscle cells by stimulating the Na+ - K+ pump. Induces apoptosis in human thymocytes.

Specifications

Color White
Description 99% by TLC
Target Proteins Phosphorylation of rb protein
CAS 50-02-2
Molecular Formula C22H29FO5
Formula Weight 392.5g/mol
Solubility DMSO, CHCl3 or EtOH
Form Solid
Quantity 100 mg