missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Deoxyribonucleic Acid, Sodium Salt, Calf Thymus

Purified DNA from calf thymus

$490.00

Specifications

CAS 73049-39-5
Color Off-white
Content And Storage Store at −20°C
Synonym DNA, Calf Thymus
Form Solid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
26181GM
View Documents Promotion Details
MilliporeSigma™
26181GM
1 g
Each for $490.00
Only null left
 
Description

Description

A260/280 nm: ≥ 1.8); hyperchromicity: ≥27%
Specifications

Specifications

73049-39-5
Store at −20°C
Solid
Off-white
DNA, Calf Thymus
Videos
SDS
Documents

Documents