Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Deoxyribonucleic Acid, Sodium Salt, Calf Thymus

Catalog No. 26181GM
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26181GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. 26181GM missing translation for 'mfr' MilliporeSigma™ missing translation for 'supplierNo' 26181GM

Purified DNA from calf thymus

A260/280 nm: ≥ 1.8); hyperchromicity: ≥27%

Specifications

CAS 73049-39-5
Color Off-white
Content And Storage Store at −20°C
Quantity 1 g
Synonym DNA, Calf Thymus
Form Solid