missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Deoxyhypusine Synthase Inhibitor, GC7

Catalog No. 25954510MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25954510MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25954510MG Supplier MilliporeSigma™ Supplier No. 25954510MG

The Deoxyhypusine Synthase Inhibitor, GC7, also referenced under CAS 150333-69-0, controls the biological activity of Deoxyhypusine Synthase.

A cell-permeable inhibitor that targets the spermidine-binding site of Deoxyhypusine Synthase (Ki = 9.7 nM), an enzyme which is responsible for the first step hypunisation of eukaryotic initiation factor 5A (eIF5A). In vivo, it demonstrates almost complete cessation of hypusine formation in CHO cells at ≥ 1μM. Several studies indicate this compound as a useful tool for the down-regulation of eIF5A, a protein essential for cell proliferation. For example, it exhibits an anti-proliferative effect on both BCR-ABL -positive and -negative leukemia cell lines (K562 and HL-60) dose-dependently, at concentrations between 5μM and 40μM. In addition, it is shown to significantly impair the growth and invasion of primary HCC cells (HepG2 and Hep3B) in the 10-20μM range, and against non-tumorigenic CL-48 cells at concentrations as high as 40μM, with no apparent cytotoxicity. At 25μM, GC7 treatment is shown to reverse eIF5a-mediated Skeletal Muscle Stem Cell Differentiation.

Specifications

Color White
Description >95% by NMR
CAS 150333-69-0
Molecular Formula C8H22N4O4S
Formula Weight 270.35g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Powder
Quantity 10 mg