missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Protease Inhibitor Cocktail Set V, EDTA-Free
A cocktail of four protease inhibitors for the inhibition of serine, cysteine, but not metalloproteases.
$522.15 - $522.15
Specifications
| Target Proteins | Serine and cysteine proteases, but not metalloproteases |
|---|---|
| Solubility | H2O |
| Content And Storage | Hygroscopic |
| Form | Lyophilized |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
53-913-710VL
|
MilliporeSigma™
53913710VL |
10 Vials |
Each for $522.15
|
|
|||||
Description
A cocktail of four protease inhibitors for the inhibition of serine, cysteine, but not metalloproteases. Reconstitute each vial with 1 ml H2O to obtain a 100X stock solution. 1X stock solution contains 500μM AEBSF, HCl (Cat. No. 101500), 150nM Aprotinin (Cat. No. 616370), 1μM E-64 (Cat. No. 324890), and 1μM Leupeptin Hemisulfate (Cat. No. 108975). Supplied with a data sheet.Specifications
| Serine and cysteine proteases, but not metalloproteases | |
| H2O | |
| Hygroscopic | |
| Lyophilized |