missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Protease Inhibitor Cocktail Set V, EDTA-Free

Catalog No. 53913710VL
Change view
Click to view available options
Quantity:
10 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53913710VL 10 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53913710VL Supplier MilliporeSigma™ Supplier No. 53913710VL

A cocktail of four protease inhibitors for the inhibition of serine, cysteine, but not metalloproteases.

A cocktail of four protease inhibitors for the inhibition of serine, cysteine, but not metalloproteases. Reconstitute each vial with 1mL H2O to obtain a 100X stock solution. 1X stock solution contains 500μM AEBSF, HCl (Cat. No. 101500), 150nM Aprotinin (Cat. No. 616370), 1μM E-64 (Cat. No. 324890), and 1μM Leupeptin Hemisulfate (Cat. No. 108975). Supplied with a data sheet.

Specifications

Target Proteins Serine and cysteine proteases, but not metalloproteases
Solubility H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 10 Vials