missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Ru360
Description
A cell-permeable oxygen-bridged dinuclear ruthenium amine complex that has been shown to bind to mitochondria with high affinity (Kd = 340 pM). Specifically blocks Ca2+ uptake into mitochondria in vitro (IC50 = 184 pM) and in situ in intact myocytes (complete block after incubation with ∽10μM of Ru360 for 30 min). Does not affect other cellular Ca2+ transport processes involved in cardiac muscle contraction, even at micromolar levels.
Specifications
Specifications
| Color | Reddish Brown |
| Target Proteins | Ca2+ uptake into mitochondria in vitro |
| Molecular Formula | C2H26Cl3N8O5Ru2 |
| Formula Weight | 550.8g/mol |
| Solubility | deoxygenated H2O |
| Content And Storage | Hygroscopic, Protect from light |
| Form | Solid |
| Quantity | 1 Set |
| Purity | 97% by capillary electrophoresis |