missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Ru360

Catalog No. 5574401SET
Change view
Click to view available options
Quantity:
1 Set
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5574401SET 1 Set
557440500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5574401SET Supplier MilliporeSigma™ Supplier No. 5574401SET

A cell-permeable oxygen-bridged dinuclear ruthenium amine complex that has been shown to bind to mitochondria with high affinity (Kd = 340 pM).

A cell-permeable oxygen-bridged dinuclear ruthenium amine complex that has been shown to bind to mitochondria with high affinity (Kd = 340 pM). Specifically blocks Ca2+ uptake into mitochondria in vitro (IC50 = 184 pM) and in situ in intact myocytes (complete block after incubation with ∽10μM of Ru360 for 30 min). Does not affect other cellular Ca2+ transport processes involved in cardiac muscle contraction, even at micromolar levels.

Specifications

Color Reddish Brown
Target Proteins Ca2+ uptake into mitochondria in vitro
Molecular Formula C2H26Cl3N8O5Ru2
Formula Weight 550.8g/mol
Solubility deoxygenated H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 1 Set
Purity 97% by capillary electrophoresis