missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Cytoplasmic Dynein Inhibitor, Ciliobrevin D
Description
Packaged Under Inert Gas
Specifications
Specifications
| Color | Orange |
| Description | 99% by HPLC |
| Product Type | Inhibitor |
| Molecular Formula | C17H8Cl3N3O2 |
| Formula Weight | 392.6g/mol |
| Solubility | DMSO |
| Content And Storage | Protect from light |
| Form | Powder |
| Quantity | 10 mg |