missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Conduritol B Epoxide

Catalog No. 234599100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
234599100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 234599100MG Supplier MilliporeSigma™ Supplier No. 234599100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Conduritol B Epoxide, CAS 6090-95-5, is an irreversible Inhibitor of glucocerebrosidase in neurons. Also inhibits α-glucosidase activity in a variety of species.

Inhibits α-glucosidase activity in mammals, snails, sweet almonds, and yeast. An irreversible, potent, and specific inhibitor of glucocerebrosidase in cultured neurons. Has also been shown to inhibit α-glucosidase from yeast and rabbit intestinal sucrase-isomaltase complex.

Specifications

Color White
Description 98% by HPLC
Product Type Inhibitor
Target Proteins α-glucosidase
CAS 6090-95-5
Molecular Formula C6H10O5
Formula Weight 162.1g/mol
Solubility DMSO or H2O
Content And Storage Protect from moisture
Form Solid
Quantity 100 mg
Show More Show Less