missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Complement C1q-Depleted Serum, Human

Catalog No. 2344011ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2344011ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2344011ML Supplier MilliporeSigma™ Supplier No. 2344011ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For detection and quantitation of hemolytically active complement components

Specifications

Content And Storage ≤ -70°C 
Form Solution
Quantity 1 mL
Tested For Negative for HBsAg and for HCV, HIV-I, and HIV-II antibodies
Formulation In PBS, ≤0.1% sodium azide, pH 7.4
Preservative No

For Research Use Only