missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ CMP-Sialic Acid, Disodium Salt InSolution™

Catalog No. 5052230001
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5052230001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5052230001 Supplier MilliporeSigma™ Supplier No. 5.05223.0001

A 25 mM (10 mg/608 μL) sterile-filtered solution of CMP-Sialic Acid, Disodium Salt in H2O.

  • Packaged Under Inert Gas
  • Deactivate with 5% Bleach

Specifications

Formulation A 25 mM (10mg/608μL) sterile-filtered solution of CMP-Sialic Acid, Disodium Saltin H2O.
Description 95% by HPLC
Molecular Formula C20H29N4O16P·2Na
Formula Weight 658.4g/mol
Content And Storage Avoid Freeze-Thaw Cycles, Protect from Light
Form Liquid
Quantity 10 mg