missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Cleland’s Reagent, ULTROL™ Grade

Protective agent for SH groups

Supplier:  MilliporeSigma™ 23315310GM

 View more versions of this product

Catalog No. 23315310GM


Only null left
Explore available promotions

Description

Description

  • Absorbance: (100mM, 280nm): ≤0.05
  • Oxidized dithiothreitol: ≤0.5%
Specifications

Specifications

154.24
12-3-3483
2°C to 8°C
10 g
EtOH, Water
≥99% by titration
C4H10O2S2
Solid
Dithiothreitol, DTT
Calbiochem Cleland’s Reagent
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions