missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ CENTA™ β-Lactamase Substrate

Catalog No. 21947525MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21947525MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21947525MG Supplier MilliporeSigma™ Supplier No. 21947525MG

CENTA™ β-Lactamase Substrate, CAS 9073-60-3, is a chromogenic β-lactamase substrate for both Gram-negative and Gram-positive bacteria.

Chromogenic β-lactamase substrate for both gram-negative and gram-positive bacteria that is comparable to other chromogens (PADAC™ Substrate and nitrocefin). Hydrolysis of the beta-lactam ring causes a color change from light yellow (λmax: 340 nm) to chrome yellow (λmax: 405 nm). Relatively unaffected by commonly used microbiological media and human serum. Stable in H2O and in the presence of cysteine, mercaptoethanol and dimercaptopropanol.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Specifications

Color Yellow
Description 98% by HPLC
Molecular Formula C21H17N3O8S3
Solubility 25 mM Phosphate Buffer or MeOH
Form Powder
Quantity 25 mg