missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Cdc42/Rac1 GTPase Inhibitor, ML141

Catalog No. 21770825MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21770825MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21770825MG Supplier MilliporeSigma™ Supplier No. 21770825MG

The Cdc42/Rac1 GTPase Inhibitor, ML141 controls the biological activity of Cdc42/Rac1 GTPase. This small molecule/inhibitor is primarily used for Membrane applications.

Packaged Under Inert Gas

Specifications

Color Yellow-white
Description 99% by HPLC
Molecular Formula C22H21N3O3S
Formula Weight 407.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 25 mg