missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Cathepsin D/E Substrate, Fluorogenic

Catalog No. 2193601MG
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2193601MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2193601MG Supplier MilliporeSigma™ Supplier No. 2193601MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An internally quenched fluorogenic 11-mer peptide that acts as a sensitive and selective substrate for cathepsins D and E, but not cathepsins B, H, or L .

  • Packaged Under Inert Gas
  • Peptide Sequence: MOCAc-Gly-Lys-Pro-Ile-Leu-Phe∼Phe-Arg-Leu-Lys(Dnp)-D-Arg-NH2MOCAc = (7-Methoxycoumarin-4-yl)acetyl

Specifications

Color Yellow
Formulation Supplied as a trifluoroacetate salt.
Description 97% by HPLC
Target Proteins Substrate for cathepsins D and E
Molecular Formula C85H122N22O19
Formula Weight 1756g/mol
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Lyophilized
Quantity 1 mg