missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Cathepsin B Substrate III, Fluorogenic

Catalog No. 21939225MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21939225MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21939225MG Supplier MilliporeSigma™ Supplier No. 21939225MG

Sensitive fluorogenic substrate for the quantitative determination of cathepsin B activity.

Peptide Sequence: Z-Arg-Arg-AMC, 2HCl

Specifications

Color White
Description 95% by HPLC
Target Proteins Cathepsin B
Molecular Formula C30H39N9O6·2HCl
Formula Weight 694.6g/mol
Solubility DMSO or H2O
Content And Storage Protect from Light, Protect from Moisture
Form Lyophilized
Quantity 25 mg