missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Capsaicin
Description
An active constituent of cayenne pepper with excitatory and desensitizing effects on a subset of primary afferent sensory neurons. An antagonist of acetylcholine-induced responses in rat PC12 cells. Exhibits anti-nociceptive and anti-inflammatory effects. Inhibits NF-κB activation by TNF. Reversibly inhibits platelet aggregation.
Specifications
Specifications
| Color | Off-white |
| Description | 98% by TLC |
| Product Type | Inhibitor |
| CAS | 404-86-4 |
| Molecular Formula | C18H27NO3 |
| Formula Weight | 305.4g/mol |
| Solubility | EtOH |
| Form | Solid |
| Quantity | 100 mg |