missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Substance P

Neuropeptide found in high concentrations in the gut.

$279.11 - $279.11

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Target Proteins Substance P receptors
CAS 33507-63-0
Molecular Formula C63H98N18O13S
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
05-230-6005MG
View Documents
MilliporeSigma™
052306005MG
5 mg
Each for $279.11
Only null left
 
Description

Description

Peptide Sequence: H-Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Gly-Leu-Met-NH2
Specifications

Specifications

White
Substance P receptors
C63H98N18O13S
5% Acetic Acid
Solid
Supplied as a trifluoroacetate salt.
33507-63-0
1347.6g/mol
Hygroscopic
96% by HPLC
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'