missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Substance P

Catalog No. 052306005MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
052306005MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 052306005MG Supplier MilliporeSigma™ Supplier No. 052306005MG

Neuropeptide found in high concentrations in the gut.

Peptide Sequence: H-Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Gly-Leu-Met-NH2

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Target Proteins Substance P receptors
CAS 33507-63-0
Molecular Formula C63H98N18O13S
Formula Weight 1347.6g/mol
Solubility 5% Acetic Acid
Content And Storage Hygroscopic
Form Solid
Quantity 5 mg
Purity 96% by HPLC
Show More Show Less