missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ CaCC Blocker IV, Benzbromarone

Catalog No. 5040410001
Change view
Click to view available options
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
5040410001 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5040410001 Supplier MilliporeSigma™ Supplier No. 5.04041.0001

A cell-permeable uricosuric agent that, in addition to its known efficacy in gout treatment, acts as a reversible CaCC blocker and is reported to be more potent than NFA (Cat

A cell-permeable uricosuric agent that, in addition to its known efficacy in gout treatment, acts as a reversible CaCC blocker and is reported to be more potent than NFA (Cat. No. 481987) and NPPB (Cat. No. 484100) in suppressing 500nM Ionomycin- (Cat. Nos. 407950, 407952, and 407953) induced I- influx in TMEM16A-expressing HEK293 cells (IC50 = 9.97, 140, and 150μM, respectively) as well as in blocking voltage-gated Cl- current in TMEM16B-expressing HEK293 cells, while exhibiting much reduced potency against ENaC (α, β, and γ) or CFTR. Shown to significantly reduce mucin secretion from NHBE (normal human bronchial epithelial) and suppress human ASM (airway smooth muscle) contraction upon muscarinic receptor stimulation.

Specifications

Color White
Description 97% by HPLC
Target Proteins TMEM16A-CaCC
Molecular Formula C17H12Br2O3
Formula Weight 424.1g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 100 mg