missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ CA-074 Me

Catalog No. 2055311MG
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2055311MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2055311MG Supplier MilliporeSigma™ Supplier No. 2055311MG

CA-074 Me, CAS 147859-80-1, is a cell-permeable analog of CA-074 that acts as an irreversible inhibitor of intracellular cathepsin B.

Cell Permeable

Specifications

Color White
Description 98% by HPLC
Target Proteins cathepsin B
Molecular Formula C19H31N3O6
Formula Weight 397.5g/mol
Solubility DMSO
Form Solid
Quantity 1 mg