missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ AEBSF, Hydrochloride
AEBSF, CAS 30827-99-7, is a specific irreversible inhibitor of chymotrypsin, kallikrein, plasmin, thrombin, trypsin, and related thrombolytic enzymes. A nontoxic alternative to PMSF and DFP.
Supplier: MilliporeSigma™ 1015001GM
Description
Packaged Under Inert GasSpecifications
| White to Off-white | |
| 30827-99-7 | |
| 239.7g/mol | |
| Protect from moisture | |
| 1 g |
| chymotrypsin | |
| C8H10FNO2S·HCl | |
| H2O | |
| Solid | |
| >97% by HPLC |