missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ (±)-Blebbistatin

Catalog No. 2033905MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2033905MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2033905MG Supplier MilliporeSigma™ Supplier No. 2033905MG

(±)-Blebbistatin, CAS 674289-55-5, is a cell-permeable, selective, and reversible inhibitor of nonmuscle myosin II. Blocks cell blebbing and disrupts cell migration & cytoKinesis in vertebrate cells.

  • Packaged Under Inert Gas
  • Cell Permeable

Specifications

Color Yellow
Description 97% by HPLC
Target Proteins ATPase
CAS 674289-55-5
Molecular Formula C18H16N2O2
Formula Weight 292.3g/mol
Solubility DMSO or MeOH
Content And Storage Protect from light
Form Solid
Quantity 5 mg