missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ (-)-Blebbistatin
Description
Packaged Under Inert Gas
Specifications
Specifications
| Color | Yellow |
| Description | 98% by Chiral HPLC |
| Product Type | Inhibitor |
| Target Proteins | ATPase |
| CAS | 856925-71-8 |
| Molecular Formula | C18H16N2O2 |
| Formula Weight | 292.3g/mol |
| Solubility | DMSO or MeOH |
| Content And Storage | Protect from light |
| Form | Solid |
| Show More |