missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Beclin-1 Activator I, TAT-Beclin-1, Scrambled

Catalog No. 5310380001
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5310380001 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5310380001 Supplier MilliporeSigma™ Supplier No. 5.31038.0001

A randomly shuffled Beclin-1 (267-284) peptide made cell-permeable by covalently attaching to HIV-1 Tat protein transduction domain via a diglycine linker

A randomly shuffled Beclin-1 (267-284) peptide made cell-permeable by covalently attaching to HIV-1 Tat protein transduction domain via a diglycine linker. Reported to serve as a suitable control for autophagy-inducing peptides (Cat. nos. 506048 and 506416).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Specifications

Color White
Formulation Supplied as trifluoroacetate salt.
Description 95% by HPLC
Molecular Formula C164H251N57O45
Solubility H2O
Content And Storage Hygroscopic, Protect from light
Form Powder
Quantity 5 mg