missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Forskolin, Coleus forskohlii

Catalog No. 34427010MG
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34427010MG 10 mg
34427025MG 25 mg
34427050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 34427010MG Supplier MilliporeSigma™ Supplier No. 34427010MG

Forskolin, CAS 66575-29-9, is a cell-permeable diterpene inotropic agent that acts as an activator of adenylate cyclase (EC50 = 4 mM).

Cell Permeable

Specifications

Color White to Off-white
Target Proteins Adenylate cyclase
CAS 66575-29-9
Molecular Formula C22H34O7
Formula Weight 410.5g/mol
Solubility DMSO
Form Crystalline Solid
Quantity 10 mg
Purity 99% by HPLC