missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Forskolin, 7-Deacetyl-7-[O-(N-methylpiperazino)-γ-butyryl]-, Dihydrochloride

Catalog No. 3442735MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3442735MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3442735MG Supplier MilliporeSigma™ Supplier No. 3442735MG

Chemically-modified forskolin with greater stability and water solubility than the parent compound.

Chemically-modified forskolin with greater stability and water solubility than the parent compound. Activates adenylate cyclase (EC50 = 13μM). Inhibits 5HT-stimulated inositol phosphate accumulation in smooth muscle cells.

Specifications

Color White
Target Proteins Adenylate cyclase
CAS 115116-37-5
Molecular Formula C29H48N2O7·2HCl
Formula Weight 609.6g/mol
Solubility MeOH or H2O
Content And Storage Hygroscopic
Form Solid
Quantity 5 mg
Purity 92% by HPLC