missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ APT1 Inhibitor, palmostatin B

Catalog No. 1785015MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1785015MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1785015MG Supplier MilliporeSigma™ Supplier No. 1785015MG

The APT1 Inhibitor, palmostatin B controls the biological activity of APT1.

Packaged Under Inert Gas

Specifications

Color Off-white
Description 95% by HPLC
Molecular Formula C23H36O4
Formula Weight 376.53g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg